Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dioxane, PHOTREX™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

123-91-1
C4H8O2
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
CHEBI:47032

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry and Liquid Scintillation Counting
Flash Point 12.00°C
Quantity 1L
CAS 123-91-1
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.03kg
Molecular Formula C4H8O2
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES C1COCCO1
IUPAC Name 1,4-dioxane
Molecular Weight (g/mol) 88.106
PubChem CID 31275
ChEBI CHEBI:47032
Formula Weight 88.11
Chemical Name or Material p-Dioxane
Grade PHOTREX™ Reagent
Show More Show Less