Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Hydroxycinnamic Acid, Predominantly trans 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
501-98-4
C9H8O3
164.16
MFCD00004399
NGSWKAQJJWESNS-ZZXKWVIFSA-N
p-coumaric acid,4-hydroxycinnamic acid,p-hydroxycinnamic acid,4-coumaric acid,trans-4-hydroxycinnamic acid,trans-p-coumaric acid,p-cumaric acid,3-4-hydroxyphenyl acrylic acid,para-coumaric acid,hydroxycinnamic acid
637542
CHEBI:32374
(E)-3-(4-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC=C1C=CC(=O)O)O
501-98-4
C9H8O3
164.16
MFCD00004399
NGSWKAQJJWESNS-ZZXKWVIFSA-N
p-coumaric acid,4-hydroxycinnamic acid,p-hydroxycinnamic acid,4-coumaric acid,trans-4-hydroxycinnamic acid,trans-p-coumaric acid,p-cumaric acid,3-4-hydroxyphenyl acrylic acid,para-coumaric acid,hydroxycinnamic acid
637542
CHEBI:32374
(E)-3-(4-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC=C1C=CC(=O)O)O
Melting Point 214°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 100g
Color Beige to White or Green
Assay Percent Range 98%
Linear Formula HOC6H4CH=CHCO2H
Beilstein 10, 297
Merck Index 14, 2560
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,practically insoluble in benzene and ligroine
Formula Weight 164.16
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material p-Hydroxycinnamic acid, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning