Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Methylstyrene, stabilized, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
500mL
2.5L
Packaging:
Glass bottle
622-97-9
C9H10
118.18
JLBJTVDPSNHSKJ-UHFFFAOYSA-N
4-methylstyrene,1-methyl-4-vinylbenzene,p-methylstyrene,4-vinyltoluene,p-vinyltoluene,benzene, 1-ethenyl-4-methyl,styrene, p-methyl,p-methyl styrene,1-p-tolylethene,para-methylstyrene
12161
1-ethenyl-4-methylbenzene
CC1=CC=C(C=C1)C=C
622-97-9
C9H10
118.18
JLBJTVDPSNHSKJ-UHFFFAOYSA-N
4-methylstyrene,1-methyl-4-vinylbenzene,p-methylstyrene,4-vinyltoluene,p-vinyltoluene,benzene, 1-ethenyl-4-methyl,styrene, p-methyl,p-methyl styrene,1-p-tolylethene,para-methylstyrene
12161
1-ethenyl-4-methylbenzene
CC1=CC=C(C=C1)C=C
Specific Gravity 0.896
Density 0.8960g/mL
Stabilizer 0.1% 3, 5-di-tert.-butylcatechol
Refractive Index 1.5410 to 1.5430
Packaging Glass bottle
Boiling Point 170.0°C to 175.0°C
Quantity 10mL
Color Colorless to Yellow
Flash Point 45°C
Infrared Spectrum Authentic
Melting Point -34.0°C
Assay Percent Range 98%
Linear Formula CH3C6H4CH=CH2
Beilstein 05,485
Solubility Information Solubility in water: insoluble
Formula Weight 118.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material p-Methylstyrene
Show More Show Less

;GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.IF SWALLOWED: Immediately call a POISON CENTER or doctor/phys