Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p-Naphtholbenzein Indicator 0.025% w/v in Acetic Acid, Reagents
Click to view available options
Quantity:
1 L
Specifications
Specifications
Chemical Name or Material | p-Naphtholbenzein Indicator 0.025% w/v in Acetic Acid |
CAS | 64-19-7, 145-50-6 |
Synonym | alpha-Naphtholbenzein, para-Naphtholbenzein, 1-Naphtholbenzein |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.05 |
Color | Orange/Red |
pH | <2 |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction