Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar P-NTRO BLUE TETZOLIUM CL 250MG

Melting Point ∽190°C (dec.)
Quantity 250mg
CAS 298-83-9
Packaging Plastic bottle
Physical Form Crystalline Powder
Molecular Formula C40H30Cl2N10O6
Formula Weight 817.64
MDL Number MFCD00012159
Synonym 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)tetrazolium dichloride
Chemical Name or Material p-Nitro Blue Tetrazolium Chloride (NBT)