Alfa Aesar™ p-Phenetidine, 98%

Manufacturer: Alfa Aesar™ A1041636

Quantity 500g

 View more versions of this product

Catalog No. AAA1041636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 156-43-4
    Molecular Formula C8H11NO
    Formula Weight 137.18
    MDL Number MFCD00007865
    UN Number UN2311
    Beilstein 606666
    Merck Index 14,7228
    Synonym 4-Ethoxyaniline; 4-Aminophenetole
    Chemical Name or Material p-Phenetidine

    Specifications

    Formula Weight 137.18
    Health Hazard 2 GHS H Statement
    H341-H302-H312-H332-H319-H317
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 156-43-4
    Synonym 4-Ethoxyaniline; 4-Aminophenetole
    Odor Unpleasant
    RTECS Number SI6465500
    Molecular Formula C8H11NO
    Melting Point 2° to 4°C
    Quantity 500g
    Flash Point 115°C (239°F)
    MDL Number MFCD00007865
    UN Number UN2311
    Beilstein 606666
    Merck Index 14,7228
    Health Hazard 1 Warning
    Boiling Point 254°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENETIDINES
    Chemical Name or Material p-Phenetidine
    Assay 98%

    Description

    Applications
    This chemical is mainly used as intermediate for several materials also used as medicine, dye, food preservatives, feed additives and rubber age-resistant intermediates.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Store in dark.

    GHS H Statement
    H341-H302-H312-H332-H319-H317
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit