Learn More
p-Phenylenediamine, 97%, Thermo Scientific Chemicals
Description
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.
Solubility
Soluble in alcohol, chloroform, ether and hot benzene.
Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Specifications
Melting Point | 138°C to 142°C |
Boiling Point | 267°C to 271°C |
Flash Point | 156°C (312°F) |
Assay Percent Range | 97% |
Quantity | 250 g |
UN Number | UN1673 |
Beilstein | 742029 |
Merck Index | 14,7285 |
Solubility Information | Soluble in alcohol,chloroform,ether and hot benzene. |
Formula Weight | 108.14 |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H317-H319 |
Hazard Statement | GHS H Statement H301-H311-H331-H319-H317 |
Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES |
EINECSNumber | 203-404-7 |
RTECSNumber | SS8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.