Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ p-Phenylenediamine dihydrochloride, 99%

Manufacturer:  Alfa Aesar™ A106380B

Quantity 1000g

 View more versions of this product

Catalog No. AAA106380B


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

624-18-0
181.07
UN2811
14,7285
p-Phenylenediamine dihydrochloride
C6H8N2·2HCl
MFCD00013002
3911749
1,4-Diaminobenzene dihydrochloride

Specifications

624-18-0
181.07
UN2811
14,7285
≈265°C
p-Phenylenediamine dihydrochloride
C6H8N2·2HCl
MFCD00013002
3911749
1,4-Diaminobenzene dihydrochloride
1000g
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : ST0350000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit