Alfa Aesar™ p-Phenylenediamine dihydrochloride, 99%

Manufacturer: Alfa Aesar™ A1063818

Quantity 50g

 View more versions of this product

Catalog No. AAA1063818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 624-18-0
    Molecular Formula C6H8N2·2HCl
    Formula Weight 181.07
    MDL Number MFCD00013002
    UN Number UN2811
    Beilstein 3911749
    Merck Index 14,7285
    Synonym 1,4-Diaminobenzene dihydrochloride
    Chemical Name or Material p-Phenylenediamine dihydrochloride

    Specifications

    Formula Weight 181.07
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,7285
    Quantity 50g
    RTECS Number ST0350000
    MDL Number MFCD00013002
    CAS 624-18-0
    Health Hazard 1 Danger
    Melting Point ≈265°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    Molecular Formula C6H8N2·2HCl
    Beilstein 3911749
    Synonym 1,4-Diaminobenzene dihydrochloride
    UN Number UN2811
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H319-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Chemical Name or Material p-Phenylenediamine dihydrochloride
    Assay 99%

    GHS H Statement
    H301-H311-H331-H319-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit