Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Selectin/CD62P Mouse anti-Human, Rat, Goat, Sheep, Alexa Fluor™ 532, Clone: Psel.KO.2.12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10065391AF532

Catalog No. N106391A32


Add to cart

Description

Description

P-Selectin/CD62P Monoclonal antibody specifically detects P-Selectin/CD62P in Human, Rat, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

P-Selectin/CD62P
Psel.KO.2.12
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
SELP
P-Selectin transfected 300.19 cells.
Protein G purified
Store at 4°C in the dark.
Monoclonal
NB100-65391 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. Clone Psel.KO.2.12 is reported to inhibit P-selectin-dependent adhesion between HL60 cells and P-selectin transfected COS cells.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
Mouse
IgG1
0.1 mL
Primary
6403.0
Goat, Human, Rat, Sheep
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit