Learn More
Novus Biologicals™ Human P-Selectin/CD62P ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP189873
Description
The Human P-Selectin/CD62P ELISA Kit (Colorimetric) measures human P-Selectin/CD62P in cell culture supernatant, serum, plasma (edta, citrate, heparin).Specifications
0.63 - 40.00ng/mL | |
ELISA Kit | |
ELISA | |
SELP | |
96 Tests | |
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) |
0.20ng/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
6403 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sP-selectin, HRP-Conjugate anti-human sP-selectin monoclonal antibody, Human sP-selectin Standard lyophilized, 80ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Storage of components varies. See protocol for specific instructions. | |
P-Selectin/CD62P |