Learn More
P-Selectin Monoclonal Antibody (HI62P), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA134004
Description
MA1-34004 detects CD62P from human samples. MA1-34004 has been successfully used in flow cytometry and Western blot applications. The MA1-34004 immunogen is: Human platelets. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
P-selectin is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate.Specifications
P-Selectin | |
Monoclonal | |
1 mg/mL | |
SELP | |
RP1-86F14.2, CD62, CD62P, FLJ45155, GMP140, GRMP, LECAM3, PADGEM, PSEL | |
Mouse | |
Protein G | |
RUO | |
6403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
HI62P | |
Unconjugated | |
P16109 | |
SELP | |
Human platelets. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.