Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLKL (p Ser345) Rabbit anti-Mouse, Clone: JM92-37, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266953
Description
MLKL (p Ser345) Monoclonal antibody specifically detects MLKL (p Ser345) in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
MLKL (p Ser345) | |
JM92-37 | |
Unconjugated | |
Purified | |
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
MLKL | |
Synthetic phospho-peptide corresponding to residues surrounding Ser345 of human MLKL | |
Monoclonal | |
Protein A purified | |
Necroptosis, Protein Kinase | |
Mouse |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
TBS (pH7.4), 1%BSA, 40% Glycerol with 0.05% Sodium Azide | |
197259 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title