Alfa Aesar™ p-Sexiphenyl, 95%

Manufacturer: Alfa Aesar™ H26586MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26586MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4499-83-6
    Molecular Formula C36H26
    Formula Weight 458.59
    MDL Number MFCD00039559
    Chemical Name or Material p-Sexiphenyl

    Specifications

    Formula Weight 458.59
    Molecular Formula C36H26
    MDL Number MFCD00039559
    CAS 4499-83-6
    Melting Point >400°C
    Quantity 250mg
    Chemical Name or Material p-Sexiphenyl
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit