Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolualdehyde 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
104-87-0
C8H8O
120.15
MFCD00006954
FXLOVSHXALFLKQ-UHFFFAOYSA-N
p-tolualdehyde,4-tolualdehyde,p-formyltoluene,p-tolylaldehyde,p-toluylaldehyde,p-methylbenzaldehyde,para-tolualdehyde,benzaldehyde, 4-methyl,para-methylbenzaldehyde,para-toluyl aldehyde
7725
CHEBI:28617
4-methylbenzaldehyde
CC1=CC=C(C=C1)C=O
104-87-0
C8H8O
120.15
MFCD00006954
FXLOVSHXALFLKQ-UHFFFAOYSA-N
p-tolualdehyde,4-tolualdehyde,p-formyltoluene,p-tolylaldehyde,p-toluylaldehyde,p-methylbenzaldehyde,para-tolualdehyde,benzaldehyde, 4-methyl,para-methylbenzaldehyde,para-toluyl aldehyde
7725
CHEBI:28617
4-methylbenzaldehyde
CC1=CC=C(C=C1)C=O
Specific Gravity 1.015
Refractive Index 1.5420 to 1.5480
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Boiling Point 204.0°C to 205.0°C
Flash Point 71°C
Melting Point -6.0°C
Quantity 2.5kg
Density 1.0150g/mL
Assay Percent Range 97%
Linear Formula CH3C6H4CHO
Beilstein 07, 297
Solubility Information Solubility in water: 0.25g/l (25°C). Other solubilities: soluble in alcohol and ether
Formula Weight 120.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material p-Tolualdehyde, 97%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 203-246-9

RTECSNumber : CU7034500

TSCA : TSCA