Alfa Aesar™ p-Tolualdehyde, 98%

Manufacturer: Alfa Aesar™ A1138436

Quantity 500g

 View more versions of this product

Catalog No. AAA1138436

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 104-87-0
    Molecular Formula C8H8O
    Formula Weight 120.15
    MDL Number MFCD00006954
    Beilstein 385772
    Synonym 4-Methylbenzaldehyde
    Chemical Name or Material p-Tolualdehyde

    Specifications

    Formula Weight 120.15
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00006954
    CAS 104-87-0
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Melting Point −6°C
    Quantity 500g
    Molecular Formula C8H8O
    Beilstein 385772
    RTECS Number CU7034500
    Flash Point 80°C (176°F)
    Boiling Point 204°C
    TSCA TSCA
    Synonym 4-Methylbenzaldehyde
    Health Hazard 1 Warning
    Chemical Name or Material p-Tolualdehyde
    Assay 98%

    Description

    Applications
    p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

    Solubility
    It is slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents

    GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit