Alfa Aesar™ p-Toluamide, 98+%

Manufacturer: Alfa Aesar™ A1155936

Quantity 500g

 View more versions of this product

Catalog No. AAA1155936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 619-55-6
    Molecular Formula C8H9NO
    Formula Weight 135.17
    MDL Number MFCD00008002
    Beilstein 774353
    Synonym 4-Methylbenzamide
    Chemical Name or Material p-Toluamide

    Specifications

    Formula Weight 135.17
    Molecular Formula C8H9NO
    CAS 619-55-6
    Melting Point 158° to 160°C
    Quantity 500g
    TSCA TSCA
    MDL Number MFCD00008002
    Beilstein 774353
    RTECS Number CV5570150
    Synonym 4-Methylbenzamide
    Chemical Name or Material p-Toluamide
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit