Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ p-Toluamide, 98+%

Manufacturer:  Alfa Aesar™ A1155914

Quantity 25g

 View more versions of this product

Catalog No. AAA1155914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

619-55-6
135.17
774353
p-Toluamide
C8H9NO
MFCD00008002
4-Methylbenzamide

Specifications

158° to 160°C
619-55-6
135.17
774353
p-Toluamide
25g
C8H9NO
MFCD00008002
4-Methylbenzamide
≥98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : CV5570150

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit