Alfa Aesar™ p-Toluenesulfonic acid monohydrate, 97%

Manufacturer: Alfa Aesar™ 3650622

Quantity 100g

 View more versions of this product

Catalog No. AA3650622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6192-52-5
    Molecular Formula CH3C6H4SO3H·H2O
    Formula Weight 190.22 (172.20 Anhydrous)
    MDL Number MFCD00142137
    UN Number UN2585
    Beilstein 3568023
    Merck Index 14,9533
    Synonym 4-Methylbenzenesulfonic acid monohydrate
    Chemical Name or Material p-Toluenesulfonic acid monohydrate

    Specifications

    Melting Point 103° to 106°C (anhy)
    Formula Weight 190.22 (172.20 Anhydrous)
    Assay Percent Range 97%
    Molecular Formula CH3C6H4SO3H·H2O
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White to pink
    Sensitivity Hygroscopic
    Beilstein 3568023
    Health Hazard 2 GHS H Statement
    H314-H335-H319
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Causes serious eye irritation.
    Odor Mild
    Physical Form Crystalline
    Health Hazard 1 Danger
    TSCA TSCA
    Flash Point 150° (302°F)
    UN Number UN2585
    Merck Index 14,9533
    Synonym 4-Methylbenzenesulfonic acid monohydrate
    Density 1.24g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
    Boiling Point 140°C (20mmHg)
    Quantity 100g
    MDL Number MFCD00142137
    RTECS Number DB7164000
    CAS 6192-52-5
    Chemical Name or Material p-Toluenesulfonic acid monohydrate
    Assay 97%

    GHS H Statement
    H314-H335-H319
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit