Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ p-Toluenesulfonic acid monohydrate, ACS, 98.5+%

Manufacturer:  Alfa Aesar™ 4400222

Quantity 100g

 View more versions of this product

Catalog No. AA4400222


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6192-52-5
190.22 (172.20 Anhydrous)
UN2585
14,9533
p-Toluenesulfonic acid monohydrate
C7H8O3S·H2O
MFCD00142137
3568023
PTSA; Tosic acid monohydrate
ACS Grade

Specifications

6192-52-5
C7H8O3S·H2O
MFCD00142137
3568023
1.24g/mL
Mild
White to pink
100g
Hygroscopic
≥98.5%
Crystalline
190.22 (172.20 Anhydrous)
UN2585
14,9533
PTSA; Tosic acid monohydrate
103° to 106°C (anhydrous)
140°C (20mmHg)
150°C (302°F)
p-Toluenesulfonic acid monohydrate
ACS Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID

RTECSNumber : DB7164000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit