Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic Acid TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
6192-52-5,67-64-1
C7H10O4S
190.21
KJIFKLIQANRMOU-UHFFFAOYSA-N
4-methylbenzene-1-sulfonic acid hydrate
O.CC1=CC=C(C=C1)S(O)(=O)=O
6192-52-5,67-64-1
C7H10O4S
190.21
KJIFKLIQANRMOU-UHFFFAOYSA-N
4-methylbenzene-1-sulfonic acid hydrate
O.CC1=CC=C(C=C1)S(O)(=O)=O

T-129, 104-15-4

Spectrum™ p-Toluenesulfonic Acid TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 0.001,0.999
UN Number UN1090
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL