Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic anhydride, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
4124-41-8
C14H14O5S2
326.39
MFCD00008548
PDVFSPNIEOYOQL-UHFFFAOYSA-N
p-toluenesulfonic anhydride,4-methylbenzenesulfonic anhydride,4-toluenesulfonic anhydride,p-toluenesulfonicanhydride,p-toluene sulfonic anhydride,toluene-p-sulphonic anhydride,4-methylphenyl sulfonyl 4-methylbenzenesulfonate,p-toluenesulphonic anhydride,toluene-4-sulfonic anhydride,benzenesulfonic acid, 4-methyl-, anhydride
77773
(4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
4124-41-8
C14H14O5S2
326.39
MFCD00008548
PDVFSPNIEOYOQL-UHFFFAOYSA-N
p-toluenesulfonic anhydride,4-methylbenzenesulfonic anhydride,4-toluenesulfonic anhydride,p-toluenesulfonicanhydride,p-toluene sulfonic anhydride,toluene-p-sulphonic anhydride,4-methylphenyl sulfonyl 4-methylbenzenesulfonate,p-toluenesulphonic anhydride,toluene-4-sulfonic anhydride,benzenesulfonic acid, 4-methyl-, anhydride
77773
(4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
Melting Point 124.0°C to 126.0°C
Color White to Gray
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Assay Percent Range 95%
Beilstein 11, IV, 376
Formula Weight 326.39
Physical Form Powder, Crystals and/or Chunks
Percent Purity 95%
Chemical Name or Material p-Toluenesulfonic anhydride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger