Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonyl chloride, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2kg
10kg
Packaging:
Glass bottle
Plastic drum
98-59-9
C7H7ClO2S
190.65
YYROPELSRYBVMQ-UHFFFAOYSA-N
tosyl chloride,p-toluenesulfonyl chloride,p-tosyl chloride,4-toluenesulfonyl chloride,p-toluenesulfochloride,4-methylbenzene-1-sulfonyl chloride,p-tolylsulfonyl chloride,p-toluenesulphonyl chloride,tosylchloride,p-toluenesulfonic acid chloride
7397
4-methylbenzenesulfonyl chloride
CC1=CC=C(C=C1)S(=O)(=O)Cl
98-59-9
C7H7ClO2S
190.65
YYROPELSRYBVMQ-UHFFFAOYSA-N
tosyl chloride,p-toluenesulfonyl chloride,p-tosyl chloride,4-toluenesulfonyl chloride,p-toluenesulfochloride,4-methylbenzene-1-sulfonyl chloride,p-tolylsulfonyl chloride,p-toluenesulphonyl chloride,tosylchloride,p-toluenesulfonic acid chloride
7397
4-methylbenzenesulfonyl chloride
CC1=CC=C(C=C1)S(=O)(=O)Cl
Color White
Flash Point 128°C
Melting Point 67.0°C to 69.0°C
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 134.0°C (10.0 mmHg)
Assay Percent Range 99+%
Linear Formula CH3C6H4SO2Cl
Beilstein 11,103
Fieser 01,1179; 03,292; 04,510; 05,676; 06,598; 08,489; 11,536; 13,286
Merck Index 15,9693
Solubility Information Solubility in water: hydrolyses. Other solubilities: hydrolysis to acid, freely soluble in alcohol, benzene and ether
Formula Weight 190.65
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material p-Toluenesulfonyl chloride
Show More Show Less