Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonyl cyanide, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
19158-51-1
C8H7NO2S
181.21
JONIMGVUGJVFQD-UHFFFAOYSA-N
tosyl cyanide,p-toluenesulfonyl cyanide,4-methylbenzenesulfonyl cyanide,p-toluenesulphonyl cyanide,p-toluene sulfonyl cyanide,p-tolylsulfonyl cyanide,p-toluenesulfonylcyanide,benzenesulfonylcyanide, 4-methyl,benzenesulfonyl cyanide, 4-methyl,4-methylphenyl sulfonylformonitrile
87946
(4-methylphenyl)sulfonylformonitrile
CC1=CC=C(C=C1)S(=O)(=O)C#N
19158-51-1
C8H7NO2S
181.21
JONIMGVUGJVFQD-UHFFFAOYSA-N
tosyl cyanide,p-toluenesulfonyl cyanide,4-methylbenzenesulfonyl cyanide,p-toluenesulphonyl cyanide,p-toluene sulfonyl cyanide,p-tolylsulfonyl cyanide,p-toluenesulfonylcyanide,benzenesulfonylcyanide, 4-methyl,benzenesulfonyl cyanide, 4-methyl,4-methylphenyl sulfonylformonitrile
87946
(4-methylphenyl)sulfonylformonitrile
CC1=CC=C(C=C1)S(=O)(=O)C#N
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 47.0°C to 50.0°C
Assay Percent Range 94% min. (HPLC)
Formula Weight 181.21
Percent Purity 95%
Chemical Name or Material p-Toluenesulfonyl cyanide

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 242-849-1