Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonyl fluoride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
455-16-3
C7H7FO2S
174.189
IZZYABADQVQHLC-UHFFFAOYSA-N
p-toluenesulfonyl fluoride,tosyl fluoride,4-toluenesulfonyl fluoride,benzenesulfonyl fluoride, 4-methyl,4-tolylsulfonyl fluoride,toluene-4-sulfonyl fluoride,p-methylbenzenesulfonyl fluoride,p-toluenesulphonyl fluoride,toluene-p-sulphonyl fluoride,p-methylphenylsulfonyl fluoride
9965
4-methylbenzenesulfonyl fluoride
CC1=CC=C(C=C1)S(=O)(=O)F
455-16-3
C7H7FO2S
174.189
IZZYABADQVQHLC-UHFFFAOYSA-N
p-toluenesulfonyl fluoride,tosyl fluoride,4-toluenesulfonyl fluoride,benzenesulfonyl fluoride, 4-methyl,4-tolylsulfonyl fluoride,toluene-4-sulfonyl fluoride,p-methylbenzenesulfonyl fluoride,p-toluenesulphonyl fluoride,toluene-p-sulphonyl fluoride,p-methylphenylsulfonyl fluoride
9965
4-methylbenzenesulfonyl fluoride
CC1=CC=C(C=C1)S(=O)(=O)F
Packaging Glass Bottle
Quantity 25g
Melting Point 40.0°C to 43.0°C
Boiling Point 112.0°C (16.0 mmHg)
Flash Point 106°C
Assay Percent Range 97%
Linear Formula CH3C6H4SO2F
Beilstein 11, II, 54
Solubility Information Solubility in water: hydrolysis.
Formula Weight 174.19
Percent Purity 97%
Chemical Name or Material p-Toluenesulfonyl fluoride, 97%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger