Learn More
p-Toluenesulfonyl fluoride, 98%, Thermo Scientific Chemicals
Description
p-Toluenesulfonyl fluoride is used for sulfonylation of organometallic reagents as a simple, one-step synthesis of sulfones. It is useful in the treatment of Alzheimer?s disease. It is used to study the DNS polymerase from spinach leaf extracts. It was used in isolation and separation of Pseudomonas aeruginosa membranes by density sucrose gradient centrifugation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
Specifications
Melting Point | 40°C to 41°C |
Boiling Point | 112°C (16 mmHg) |
Flash Point | 105°C (221°F) |
Quantity | 25 g |
UN Number | UN3261 |
Beilstein | 2208423 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 174.19 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H314 |
Hazard Statement | GHS H Statement H301-H314-H318 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 207-238-6 |
RTECSNumber | XT8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.