Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methylbenzenesulphonylhydrazide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4,3-c]pyridines and 1,2,3-selenadiazole derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4′,3′-c]pyridines and 1,2,3-selenadiazole derivatives.
Solubility
Soluble in water, alcohol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4′,3′-c]pyridines and 1,2,3-selenadiazole derivatives.
Solubility
Soluble in water, alcohol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 108°C to 111°C |
Density | 1.4 |
Quantity | 25 g |
UN Number | UN3226 |
Beilstein | 610130 |
Solubility Information | Soluble in water,alcohol. |
Formula Weight | 186.23 |
Percent Purity | 98% |
Chemical Name or Material | p-Toluenesulfonyl hydrazide |
Safety and Handling
Hazard Category | H242-H301-H319 |
Hazard Statement | GHS H Statement H242-H228-H301 Heating may cause a fire. Flammable solid. Toxic if swallowed. |
Precautionary Statement | P210-P220-P234-P264b-P270-P280-P301+P310-P305+P351+P338-P330-P370+P378q-P375-P380-P501c |
DOTInformation | Hazard Class: 4.1 |
EINECSNumber | 216-407-3 |
RTECSNumber | MW0210000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction