Learn More
p-Toluenesulfonyl isocyanate, 95%, Thermo Scientific Chemicals
Description
It is a reagent used to prepare acetylated syn-1,2-diols,oxazolidin-2-ones, 2,3-diamino acids, and N-tosylcarbonamides. -Toluenesulfonyl isocyanate has been used as derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS. It is also employed in the derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC and as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a reagent used to prepare acetylated syn-1,2-diols,oxazolidin-2-ones, 2,3-diamino acids, and N-tosylcarbonamides. -Toluenesulfonyl isocyanate has been used as derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-MS/MS. It is also employed in the derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC and as reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and N-tosylcarbonamides.
Solubility
Reacts with water.
Notes
Keep away from alcohols, strong bases, amines, strong oxidizing agents.
Specifications
Melting Point | 5°C |
Density | 1.295 |
Boiling Point | 144°C (10 mmHg) |
Flash Point | 145°C (293°F) |
Odor | Irritating |
Refractive Index | 1.535 |
Quantity | 25 g |
UN Number | UN2206 |
Beilstein | 391287 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H315-H319-H334-H335-H336 |
Hazard Statement | GHS H Statement H334-H335-H315-H319 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P331-P332+P313-P342+P311-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S. |
EINECSNumber | 223-810-8 |
RTECSNumber | DB9032000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.