Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonylacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
5697-44-9
C9H9NO2S
195.236
MFCD00039487
BBNNLJMGPASZPD-UHFFFAOYSA-N
p-toluenesulfonylacetonitrile,2-tosylacetonitrile,tosylacetonitrile,p-tolylsulfonylacetonitrile,4-methylphenyl sulfonyl acetonitrile,4-toluenesulfonylacetonitrile,4-methylphenyl sulfonylacetonitrile,4-toluenesulphonylacetonitrile,p-cyanomethylsulfonyl toluene,tosylmethyl isocyanide
79776
2-(4-methylphenyl)sulfonylacetonitrile
CC1=CC=C(C=C1)S(=O)(=O)CC#N
5697-44-9
C9H9NO2S
195.236
MFCD00039487
BBNNLJMGPASZPD-UHFFFAOYSA-N
p-toluenesulfonylacetonitrile,2-tosylacetonitrile,tosylacetonitrile,p-tolylsulfonylacetonitrile,4-methylphenyl sulfonyl acetonitrile,4-toluenesulfonylacetonitrile,4-methylphenyl sulfonylacetonitrile,4-toluenesulphonylacetonitrile,p-cyanomethylsulfonyl toluene,tosylmethyl isocyanide
79776
2-(4-methylphenyl)sulfonylacetonitrile
CC1=CC=C(C=C1)S(=O)(=O)CC#N
Melting Point 147°C to 151°C
Color White
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 98%
Formula Weight 195.24
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material p-Toluenesulfonylacetonitrile, 98%

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 227-173-7