Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenethiol 98%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
2.5kg
Packaging:
Glass bottle
106-45-6
C7H8S
124.2
MFCD00004851
WLHCBQAPPJAULW-UHFFFAOYSA-N
p-toluenethiol,p-thiocresol,4-thiocresol,4-methylthiophenol,p-methylthiophenol,benzenethiol, 4-methyl,4-toluenethiol,p-tolyl mercaptan,p-thiolcresol,p-tolylthiol
7811
4-methylbenzenethiol
CC1=CC=C(C=C1)S
106-45-6
C7H8S
124.2
MFCD00004851
WLHCBQAPPJAULW-UHFFFAOYSA-N
p-toluenethiol,p-thiocresol,4-thiocresol,4-methylthiophenol,p-methylthiophenol,benzenethiol, 4-methyl,4-toluenethiol,p-tolyl mercaptan,p-thiolcresol,p-tolylthiol
7811
4-methylbenzenethiol
CC1=CC=C(C=C1)S
Infrared Spectrum Authentic
Quantity 10g
Melting Point 40.0°C to 44.0°C
Boiling Point 195.0°C
Color White to Yellow
Packaging Glass bottle
Flash Point 68°C
Assay Percent Range 98%
Linear Formula CH3C6H4SH
Beilstein 06, 416
Merck Index 15, 9478
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 124.2
Physical Form Fused Solid
Percent Purity 98%
Chemical Name or Material p-Toluenethiol, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-399-1

RTECSNumber : XT8925000

TSCA : TSCA