Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Plastic Drum
Plastic bottle
99-94-5
C8H8O2
136.15
MFCD00002565
LPNBBFKOUUSUDB-UHFFFAOYSA-N
p-toluic acid,4-toluic acid,crithminic acid,p-methylbenzoic acid,p-toluylic acid,benzoic acid, 4-methyl,p-carboxytoluene,para-toluic acid,4-methyl-benzoic acid,toluenecarboxylic acid
7470
CHEBI:36635
4-methylbenzoic acid
CC1=CC=C(C=C1)C(=O)O
99-94-5
C8H8O2
136.15
MFCD00002565
LPNBBFKOUUSUDB-UHFFFAOYSA-N
p-toluic acid,4-toluic acid,crithminic acid,p-methylbenzoic acid,p-toluylic acid,benzoic acid, 4-methyl,p-carboxytoluene,para-toluic acid,4-methyl-benzoic acid,toluenecarboxylic acid
7470
CHEBI:36635
4-methylbenzoic acid
CC1=CC=C(C=C1)C(=O)O
Packaging Plastic Drum
Color Cream-Yellow to White
Quantity 5kg
Infrared Spectrum Authentic
Boiling Point 274.0°C to 275.0°C
Melting Point 179.0°C to 182.0°C
Assay Percent Range 98%
Linear Formula CH3C6H4CO2H
Beilstein 09,483
Merck Index 14,9535
Solubility Information Solubility in water: 11.6g/L (98°C). Other solubilities: soluble in alcohol and ether
Formula Weight 136.15
Physical Form Powder
Percent Purity 98%
Chemical Name or Material p-Toluic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-803-3

RTECSNumber : XU1575000

TSCA : TSCA