Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluic Hydrazide 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3619-22-5
C8H10N2O
150.181
MFCD00007607
MFFVZXOPRXMVET-UHFFFAOYSA-N
p-toluic hydrazide,p-toluic acid hydrazide,p-toluoylhydrazine,4-methylbenzoylhydrazine,p-toluic acid, hydrazide,4-methylbenzoic acid hydrazide,p-methylbenzohydrazide,p-toluohydrazide,p-methylbenzoylhydrazine,4-toluic acid hydrazide
77174
4-methylbenzohydrazide
CC1=CC=C(C=C1)C(=O)NN
3619-22-5
C8H10N2O
150.181
MFCD00007607
MFFVZXOPRXMVET-UHFFFAOYSA-N
p-toluic hydrazide,p-toluic acid hydrazide,p-toluoylhydrazine,4-methylbenzoylhydrazine,p-toluic acid, hydrazide,4-methylbenzoic acid hydrazide,p-methylbenzohydrazide,p-toluohydrazide,p-methylbenzoylhydrazine,4-toluic acid hydrazide
77174
4-methylbenzohydrazide
CC1=CC=C(C=C1)C(=O)NN
Color White to Brown
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 114°C to 118°C
Assay Percent Range 98%
Linear Formula CH3C6H4CONHNH2
Beilstein 09, 494
Formula Weight 150.18
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material p-Toluic hydrazide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 222-815-2