Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ p-Toluidine, 99+%

Click to view available options
Quantity:
250g
1000g
5000g
106-49-0
C7H9N
107.156
MFCD00007906
RZXMPPFPUUCRFN-UHFFFAOYSA-N
p-toluidine,4-toluidine,p-tolylamine,4-aminotoluene,benzenamine, 4-methyl,4-methylbenzenamine,p-methylaniline,p-methylbenzenamine,p-toluidin,p-aminotoluene
7813
CHEBI:37825
4-methylaniline
CC1=CC=C(C=C1)N
106-49-0
C7H9N
107.156
MFCD00007906
RZXMPPFPUUCRFN-UHFFFAOYSA-N
p-toluidine,4-toluidine,p-tolylamine,4-aminotoluene,benzenamine, 4-methyl,4-methylbenzenamine,p-methylaniline,p-methylbenzenamine,p-toluidin,p-aminotoluene
7813
CHEBI:37825
4-methylaniline
CC1=CC=C(C=C1)N
Applications
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

Solubility
Soluble in ethanol, pyridine, diethyl ether, acetone, carbon tetrachloride, methanol, carbon disulfide, oils and dilute acids.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Density 1.05
Sensitivity Air and light sensitive
Quantity 250g
Boiling Point 199°C to 202°C
Flash Point 88°C (190°F)
Melting Point 42°C to 46°C
UN Number UN3451
Beilstein 471281
Merck Index 14,9536
Solubility Information Soluble in ethanol,pyridine,diethyl ether,acetone,carbon tetrachloride,methanol,carbon disulfide,oils and dilute acids.
Formula Weight 107.16
Percent Purity ≥99%
Chemical Name or Material p-Toluidine
Show More Show Less

GHS H Statement
H301-H311-H331-H351-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H319-H351

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, SOLID

EINECSNumber : 203-403-1

RTECSNumber : XU3150000

TSCA : Yes

Recommended Storage : Ambient temperatures