Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number aaa1558122
Click to view available options
Quantity:
100 g
500 g
2500 g
874-60-2
C8H7ClO
154.593
MFCD00000696
NQUVCRCCRXRJCK-UHFFFAOYSA-N
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
13405
4-methylbenzoyl chloride
CC1=CC=C(C=C1)C(=O)Cl
874-60-2
C8H7ClO
154.593
MFCD00000696
NQUVCRCCRXRJCK-UHFFFAOYSA-N
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
13405
4-methylbenzoyl chloride
CC1=CC=C(C=C1)C(=O)Cl

p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

Solubility
Reacts with water alcohol.

Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
TRUSTED_SUSTAINABILITY
Melting Point °C to 2°C
Density 1.169
Boiling Point 225°C to 227°C
Flash Point 82°C (179°F)
Assay Percent Range 99%
Refractive Index 1.553
Quantity 100 g
UN Number UN3265
Beilstein 471492
Sensitivity Moisture sensitive
Solubility Information Reacts with water alcohol.
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material p-Toluoyl Chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-864-8
RTECSNumber XV1660000
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit