Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ p-Toluoylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ A1095014

Quantity 25g

 View more versions of this product

Catalog No. AAA1095014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7391-28-8
159.19
742923
p-Toluoylacetonitrile
C10H9NO
MFCD00067895
4-Methylbenzoylacetonitrile

Specifications

99° to 104°C
7391-28-8
159.19
742923
p-Toluoylacetonitrile
25g
C10H9NO
MFCD00067895
4-Methylbenzoylacetonitrile
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit