Learn More
p-Tolyl isothiocyanate, 97%, Thermo Scientific Chemicals
Description
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.
Solubility
Decompose in water.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
Specifications
Melting Point | 23°C to 26°C |
Boiling Point | 236°C to 238°C |
Flash Point | 107°C (224°F) |
Odor | Pungent |
Refractive Index | 1.6345 |
Quantity | 100 g |
UN Number | UN2923 |
Beilstein | 386032 |
Sensitivity | Moisture sensitive |
Solubility Information | Decompose in water. |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H315-H317-H319-H334-H335 |
Hazard Statement | GHS H Statement H314-H318-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: II |
EINECSNumber | 210-745-5 |
RTECSNumber | NX9500000 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.