Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolylacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
622-47-9
C9H10O2
150.18
MFCD00004353
GXXXUZIRGXYDFP-UHFFFAOYSA-N
p-tolylacetic acid,4-methylphenylacetic acid,2-p-tolyl acetic acid,2-4-methylphenyl acetic acid,p-methylphenylacetic acid,4-tolylacetic acid,4-methylphenyl acetic acid,benzeneacetic acid, 4-methyl,acetic acid, p-tolyl,2-p-tolylacetic acid
248474
2-(4-methylphenyl)acetic acid
CC1=CC=C(C=C1)CC(=O)O
622-47-9
C9H10O2
150.18
MFCD00004353
GXXXUZIRGXYDFP-UHFFFAOYSA-N
p-tolylacetic acid,4-methylphenylacetic acid,2-p-tolyl acetic acid,2-4-methylphenyl acetic acid,p-methylphenylacetic acid,4-tolylacetic acid,4-methylphenyl acetic acid,benzeneacetic acid, 4-methyl,acetic acid, p-tolyl,2-p-tolylacetic acid
248474
2-(4-methylphenyl)acetic acid
CC1=CC=C(C=C1)CC(=O)O
Boiling Point 265°C to 267°C
Packaging Glass bottle
Melting Point 90°C to 93°C
Infrared Spectrum Authentic
Color White
Quantity 100g
Assay Percent Range 99%
Linear Formula CH3C6H4CH2CO2H
Beilstein 09, 530
Formula Weight 150.18
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material p-Tolylacetic acid, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 210-738-7

RTECSNumber : AJ7569000