Alfa Aesar™ p-Tolylacetonitrile, 98+%

Manufacturer: Alfa Aesar™ A1216922

Quantity 100g

 View more versions of this product

Catalog No. AAA1216922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2947-61-7
    Molecular Formula C9H9N
    Formula Weight 131.18
    MDL Number MFCD00001922
    UN Number UN3276
    Beilstein 1099645
    Synonym 4-Methylbenzyl cyanide; 4-Methylphenylacetonitrile
    Chemical Name or Material p-Tolylacetonitrile

    Specifications

    Formula Weight 131.18
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Flash Point 106°C (222°F)
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C9H9N
    MDL Number MFCD00001922
    CAS 2947-61-7
    Beilstein 1099645
    Health Hazard 1 Danger
    Quantity 100g
    UN Number UN3276
    Synonym 4-Methylbenzyl cyanide; 4-Methylphenylacetonitrile
    Boiling Point 242° to 243°C
    Melting Point 18°C
    Chemical Name or Material p-Tolylacetonitrile
    Assay ≥98%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit