Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolylhydrazine Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
637-60-5
C7H11ClN2
158.629
MFCD00012940
HMHWNJGOHUYVMD-UHFFFAOYSA-N
p-tolylhydrazine hydrochloride,4-methylphenylhydrazine hydrochloride,4-methylphenyl hydrazine hydrochloride,4-tolylhydrazine hydrochloride,hydrazine, 4-methylphenyl-, monohydrochloride,p-tolylhydrazinium 1+ chloride,hydrazine, p-tolyl-, hydrochloride,4-methylphenylhydrazine hcl,1-4-methylphenyl hydrazine hydrochloride,p-tolyl-hydrazine hydrochloride
12504
(4-methylphenyl)hydrazine;hydrochloride
CC1=CC=C(C=C1)NN.Cl
637-60-5
C7H11ClN2
158.629
MFCD00012940
HMHWNJGOHUYVMD-UHFFFAOYSA-N
p-tolylhydrazine hydrochloride,4-methylphenylhydrazine hydrochloride,4-methylphenyl hydrazine hydrochloride,4-tolylhydrazine hydrochloride,hydrazine, 4-methylphenyl-, monohydrochloride,p-tolylhydrazinium 1+ chloride,hydrazine, p-tolyl-, hydrochloride,4-methylphenylhydrazine hcl,1-4-methylphenyl hydrazine hydrochloride,p-tolyl-hydrazine hydrochloride
12504
(4-methylphenyl)hydrazine;hydrochloride
CC1=CC=C(C=C1)NN.Cl
Color Undesignated
Quantity 250g
UN Number 2811
Formula Weight 158.63
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material p-Tolylhydrazine Hydrochloride

EINECSNumber : (3)-0468

RTECSNumber : MW0195000

TSCA : Yes