Alfa Aesar™ p-Tolylurea, 98+%

Manufacturer: Alfa Aesar™ A1115022

Quantity 100g

 View more versions of this product

Catalog No. AAA1115022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 622-51-5
    Molecular Formula C8H10N2O
    Formula Weight 150.18
    MDL Number MFCD00025433
    Beilstein 2208088
    Synonym 4-Methylphenylurea
    Chemical Name or Material p-Tolylurea

    Specifications

    Formula Weight 150.18
    Molecular Formula C8H10N2O
    MDL Number MFCD00025433
    CAS 622-51-5
    Beilstein 2208088
    RTECS Number YU4200000
    Melting Point 177° to 184°C
    Quantity 100g
    Synonym 4-Methylphenylurea
    Chemical Name or Material p-Tolylurea
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit