Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene (Certified), Fisher Chemical™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
500mL
4L
Packaging:
Glass Bottle
106-42-3
C8H10
106.168
MFCD00008556
URLKBWYHVLBVBO-UHFFFAOYSA-N
p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene
7809
CHEBI:27417
1,4-xylene
CC1=CC=C(C=C1)C
106-42-3
C8H10
106.168
MFCD00008556
URLKBWYHVLBVBO-UHFFFAOYSA-N
p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene
7809
CHEBI:27417
1,4-xylene
CC1=CC=C(C=C1)C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Linear Formula C6H4(CH3)2
Formula Weight 106.17
Physical Form Liquid
Percent Purity ≥99%
Grade Certified
Density 0.866g/cm³
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 8mbar at 20°C
Viscosity 0.648 mPa/s at 20°C
Alpha Vector Xylene
Boiling Point 138°C
Color Colorless
Melting Point 13°C
Quantity 500mL
Chemical Name or Material p-Xylene
Show More Show Less

DOTInformation : DOT Class 3, : Flammable Liquid