Alfa Aesar™ p-Xylene-d{10}, 99%(Isotopic)

Manufacturer: Alfa Aesar™ 3647306

Quantity 5g

 View more versions of this product

Catalog No. AA3647306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41051-88-1
    Formula Weight 116.25
    MDL Number MFCD00037699
    UN Number UN1307
    Merck Index 14,10081
    Chemical Name or Material p-Xylene-d{10}, Isotopic

    Specifications

    Formula Weight 116.25
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H311-H226-H332-H315
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Sensitivity Moisture Sensitive
    Melting Point 12° to 13°C
    Physical Form Liquid
    Quantity 5g
    MDL Number MFCD00037699
    CAS 41051-88-1
    UN Number UN1307
    Merck Index 14,10081
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
    Density 0.948g/mL
    Boiling Point 137° to 138°C
    Health Hazard 1 Danger
    Chemical Name or Material p-Xylene-d{10}, Isotopic
    Assay 99%

    GHS H Statement
    H311-H226-H332-H315
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit