Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene, Reagent, 98%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
106-42-3
C8H10
106.17
URLKBWYHVLBVBO-UHFFFAOYSA-N
1,4-xylene
CC1=CC=C(C)C=C1
106-42-3
C8H10
106.17
URLKBWYHVLBVBO-UHFFFAOYSA-N
1,4-xylene
CC1=CC=C(C)C=C1

X1020, 106-42-3, C8H10

Spectrum™ p-Xylene, Reagent is a highly flammable aromatic hydrocarbon that is used mainly as a solvent. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

CAS Max % 1
Percent Purity 98%
Grade Reagent
Packaging Steel Can
Refractive Index 1.495-1.497
Quantity 20 L