Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p114RhoGEF Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: 1972, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243546G

Catalog No. NBP243546G


Add to cart

Description

Description

p114RhoGEF Monoclonal antibody specifically detects p114RhoGEF in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

p114RhoGEF
1972
Western Blot
Purified
ARHGEF18
Recombinant protein encompassing a sequence within the center region of human ARHGEF18. The exact sequence is proprietary.
Protein G purified
Signal Transduction
Primary
23370
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
114 kDa Rho-specific guanine nucleotide exchange factor, EC 3.4.24, EC 3.6.1, KIAA0521rho guanine nucleotide exchange factor 18, MGC15913, p114RhoGEF, P114-RhoGEF, P114-RHO-GEF, Rho/Rac guanine nucleotide exchange factor (GEF) 18, Rho/Rac guanine nucleotide exchange factor 18, Rho-specific guanine nucleotide exchange factor p114, SA-RhoGEF, Septin-associated RhoGEF
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit