Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ PEth 16:0/18:2 Solution, 1.0 mg/mL in Methanol (as free phosphate), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco P1151ML
Description
PEth 16:0/18:2, also known as phosphatidylethanol 16:0/18:2, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/18:2, in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:2, levels is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:2 has also been investigated as a biomarker to confirm prenatal alcohol exposure (PAE).Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
942.42 | |
1 mL |
C39H72O8P · NC16H36 | |
1.0 mg/mL in methanol (as free phosphate) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Recommended Storage : -70°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction