Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120-catenin Mouse anti-Human, Rat, Allophycocyanin/Cy7, Clone: 25a, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254411APCCY7

Catalog No. N254411APC7


Add to cart

Description

Description

p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

p120-catenin
25a
Flow Cytometry
Conjugated, Purified
CTNND1
Mouse p120 Catenin (pY96)
Protein A or G purified
Phospho Specific, Signal Transduction
Primary
1500.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit