Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p120-catenin Antibody (25a), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP25335720UG
Description
p120-catenin Monoclonal specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
p120-catenin | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
Mouse | |
120 kDa | |
20 μg | |
Phospho Specific, Signal Transduction | |
1500 | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
25a | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
CTNND1 | |
Mouse p120 Catenin (pY96) | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction