Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120-catenin Rabbit anti-Human, Clone: 4383R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen p120-catenin
Clone 4383R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123078
View Documents
Novus Biologicals
NBP30780220UG
20 μg
Each for $159.82
Add to cart
 
NB123077
View Documents
Novus Biologicals
NBP30780210UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

p120-catenin Monoclonal antibody specifically detects p120-catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

p120-catenin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Synthetic peptide corresponding to p120-catenin residues within aa900-1000 of p120-catenin was used as an immunogen.
Monoclonal
Protein A or G purified
4383R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Phospho Specific, Signal Transduction
10 mM PBS with 0.05% BSA
1500
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit