Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120-catenin Rabbit anti-Human, Clone: EP66, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP276321

Catalog No. NBP276321

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

p120-catenin Monoclonal antibody specifically detects p120-catenin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

p120-catenin
EP66
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:100
Affinity Purified
CTNND1
A synthetic peptide corresponding to residues in human p120 Catenin protein
Affinity purified
Store at 4°C.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
10 mM PBS, pH 7.5, containing less than 0.1% sodium azide with 0.05% Sodium Azide
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Rabbit
IgG
0.1 mL
Primary
1500.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit