Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p130 Mouse anti-Human, Clone: KAB40, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266635
Description
p130 Monoclonal antibody specifically detects p130 in Human samples. It is validated for Western Blot.Specifications
p130 | |
KAB40 | |
Western Blot | |
PBS with 0.08% Sodium Azide | |
5934 | |
Mouse | |
IgG1 | |
Primary | |
100μg | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Purified | |
FLJ26459, P130, PRB2, RB2, RBR-2, retinoblastoma-like 2 (p130), retinoblastoma-like protein 2,130 kDa retinoblastoma-associated protein, Retinoblastoma-related protein 2 | |
RBL2 | |
Recombinant protein to the N terminus of p130 | |
Monoclonal | |
Protein A or G purified | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
Human,Mouse,Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title